Learn More
CD4 Mouse anti-Human, Clone: HP2/6 (OC-515), Abnova™
Mouse Monoclonal Antibody
Manufacturer:  Abnova Corporation MAB13735
Catalog No. 89-033-800
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Mouse monoclonal antibody raised against human CD4.
  • High specificity
  • High reactivity
Specifications
Specifications
CD4
HP2/6
Mouse monoclonal antibody raised against human CD4.
In PBS, pH 7.4 (protein stabilizer, 0.09% sodium azide).
Liquid
CD4
T cells from leukemic HPB-ALL.
Protein A/G purification
RUO
Primary
920
Flow Cytometry
Unconjugated
Flow Cytometry (5 uL/106 cells) The optimal working dilution should be determined by the end user.
P01730
CD4mut
Mouse
IgG2a
100 Reactions
Store in the dark at 4°C. Avoid prolonged exposure to light.
Monoclonal
Human
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)