Learn More
CD4 Mouse anti-Human, Cynomolgus Monkey, Rhesus Macaque, Alexa Fluor™ 594, Clone: MT310, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP252706AF594
Catalog No. NP22706A94
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
CD4 Monoclonal antibody specifically detects CD4 in Human,Cynomolgus Monkey,Rhesus Macaque samples. It is validated for Western Blot,Flow Cytometry,Immunohistochemistry,Immunohistochemistry (Paraffin),Immunofluorescence.
Specifications
Specifications
CD4
MT310
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence
CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
mt310 binds to the extracellular domain D1 of CD4 -the same site as the HIV-1 gp120-binding site
Flow Cytometry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
CD4
CD4
0.1 mL
Primary
920
Cynomolgus Monkey, Human, Rhesus Macaque