Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD4 Mouse anti-Human, Monkey, Biotin, Clone: OTI5D9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270357B

Catalog No. NBP270357B


Add to cart

Description

Description

CD4 Monoclonal antibody specifically detects CD4 in Human, Monkey samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

CD4
OTI5D9
Western Blot, Immunohistochemistry
Affinity Purified, Conjugated
CD4
Full length recombinant protein of human CD4 (NP_000607) produced in HEK293T cells.
Immunogen affinity purified
Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Innate Immunity, Regulatory Immunology, Stem Cell Markers
Primary
920.0
Immunohistochemistry, Western Blot
Biotin
PBS with 0.05% Sodium Azide
CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4
Mouse
IgG2a
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human, Monkey
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit