Learn More
CD4 Mouse anti-Human, PE-CF594, Clone: SK3 (also known as Leu3a), BD
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 566317
Catalog No. BDB566317
Add to cart
Description
Description
The SK3 monoclonal antibody specifically binds to CD4. The CD4 antigen is a 55 kDa type 1 transmembrane glycoprotein that belongs to the immunoglobulin superfamily. It is expressed on T-helper/inducer lymphocytes and monocytes. CD3+CD4+ T cells comprise approximately 28% to 58% of normal peripheral blood lymphocytes. CD4 is also expressed on 80% to 95% of normal thymocytes. The CD4 antigen is present in low density on the cell surface of monocytes and in the cytoplasm of monocytes and macrophages (CD3-CD4+). CD4 serves as a T cell coreceptor for MHC class II antigen recognition and as a receptor for the human immunodeficiency virus (HIV). The SK3 antibody inhibits HIV binding to CD4+ cells.
This antibody is conjugated to BD Horizon PE-CF594, which has been developed exclusively by BD Biosciences as a better alternative to PE-Texas Red™. PE-CF594 excites and emits at similar wavelengths to PE-Texas Red™ yet exhibits improved brightness and spectral characteristics. Due to PE having maximal absorption peaks at 496 nm and 564 nm, PE-CF594 can be excited by the blue (488nm), green (532nm) and yellow-green (561nm) lasers and can be detected with the same filter set as PE-Texas Red™ (eg, 610/20nm filter). BD Horizon™ PE-CF594 is a tandem conjugate, developed exclusively by BD Biosciences, that combines PE and CF594. PE-CF594 is a brighter alternative to PE-Texas Red™, with improved spectral characteristics. PE-CF594 reagents exhibit very consistent spillover values lot to lot, making them an ideal choice for the PE-Texas Red™ detector (610/20 nm).
Specifications
Specifications
CD4
PE-CF594
Affinity Purified
Mouse
Mouse BALB/c IgG1, κ
25 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
SK3 (also known as Leu3a)
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
L3T4, Leu3a, T-cell surface antigen T4/Leu-3, W3/25, CD4 antigen (p55)
Human Peripheral Blood T Cells
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ PE-CF594 under optimum conditions, and unconjugated antibody and free PE-CF594 were removed.
RUO
Primary
Human