Learn More
CD4 Mouse anti-Human, PE-Cy5, Clone: 4AT4, Abnova™
Mouse monoclonal antibody raised against human CD4.
Manufacturer:  Abnova Corporation MAB12375
Catalog No. 89-933-800
Add to cart
Description
Description
Specifications
Specifications
CD4
4AT4
Mouse monoclonal antibody raised against human CD4.
In PBS, pH 7.4 (0.09% sodium azide, 0.2% BSA)
CD4mut
Mouse
IgG2b
50 Reactions
Store at 4°C. Do not freeze.
Monoclonal
Human
Flow Cytometry, Immunofluorescence
PE-Cyanine5
Flow cytometry Immunofluorescence The optimal working dilution should be determined by the end user.
Liquid
CD4
CD4
Protein A/G affinity chromatography
RUO
Primary
920