Learn More
CD4 Mouse anti-Human, PerCP-Cy5.5, Clone: SK3 (also known as Leu3a), BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 566316
Description
The SK3 monoclonal antibody specifically binds to CD4. The CD4 antigen is a 55 kDa type 1 transmembrane glycoprotein that belongs to the immunoglobulin superfamily. It is expressed on T-helper/inducer lymphocytes and monocytes. CD3+CD4+ T cells comprise approximately 28% to 58% of normal peripheral blood lymphocytes. CD4 is also expressed on 80% to 95% of normal thymocytes. The CD4 antigen is present in low density on the cell surface of monocytes and in the cytoplasm of monocytes and macrophages (CD3-CD4+). CD4 serves as a T cell coreceptor for MHC class II antigen recognition and as a receptor for the human immunodeficiency virus (HIV). The SK3 antibody inhibits HIV binding to CD4+ cells.
PerCP-Cy™5.5 is a tandem conjugate that combines PerCP with a cyanine dye. PerCP-Cy5.5 is not subject to photobeaching like PerCP and can be used with stream-in-air flow cytometers. It has less Fc receptor–mediated nonspecific staining than PE-Cy5. Additionally, the PerCP-Cy5.5 tandem conjugate is not as susceptible to fixative or light instability compared to APC-Cy7 and PE-Cy7.
Specifications
CD4 | |
SK3 (also known as Leu3a) | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |
Monoclonal | |
PerCP-Cyanine5.5 | |
L3T4, Leu3a, T-cell surface antigen T4/Leu-3, W3/25, CD4 antigen (p55) | |
Human Peripheral Blood T Cells | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.