Learn More
CD4 Rabbit anti-Human, Cynomolgus Monkey, Rhesus Macaque, Alexa Fluor™ 350, Clone: MT310, Novus Biologicals™
Rabbit Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP252663AF350
Catalog No. NP22663A30
Add to cart
Description
Description
CD4 Monoclonal antibody specifically detects CD4 in Human,Cynomolgus Monkey,Rhesus Macaque samples. It is validated for Western Blot,Flow Cytometry,Immunohistochemistry,Immunohistochemistry (Paraffin),Immunofluorescence.
Specifications
Specifications
CD4
MT310
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence
CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4
Rabbit
IgG Kappa
Store at 4°C in the dark.
Monoclonal
mt310 binds to the extracellular domain D1 of CD4 -the same site as the HIV-1 gp120-binding site
Flow Cytometry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
CD4
CD4
0.1 mL
Primary
920
Cynomolgus Monkey, Human, Rhesus Macaque