Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD4 Rabbit anti-Human, Janelia Fluor 549, Clone: 13B8.2, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP252670JF549

Catalog No. NBP52670549


Add to cart

Description

Description

CD4 Monoclonal antibody specifically detects CD4 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications

Specifications

CD4
13B8.2
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Purified
CD4
Human HSB2-T cells.
Protein A or G purified
Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Innate Immunity, Regulatory Immunology, Stem Cell Markers
Primary
920
Human
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4
Rabbit
IgG Kappa
0.1mL
Store at 4°C in the dark.
Monoclonal
Recognises human CD4.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit