Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD4, Rat anti-Human, PE-Cy7, Clone: YNB46.1.8 (Campath-9H), Novus Biologicals
Rat Monoclonal Antibody
Manufacturer: Novus Biologicals NBP275900PECY7100UL
Description
CD4 Monoclonal antibody specifically detects CD4 in Human Samples. It is validated for Flow Cytometry.Specifications
CD4 | |
YNB46.1.8 (Campath-9H) | |
Flow Cytometry | |
Purified | |
CD4 | |
Rat T-cell line (NB2) transfected with the human CD4 | |
Protein A purified | |
RUO | |
Store at 4°C in the dark. Do not freeze. | |
Monoclonal | |
Recognizes the 55kDa human CD4 cell surface antigen, expressed by helper T lymphocytes and by monocytes. |
Flow Cytometry | |
PE-Cyanine7 | |
PBS | |
CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4 | |
Rat | |
IgG1 Kappa | |
100 uL | |
Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Innate Immunity, Regulatory Immunology, Stem Cell Markers | |
Primary | |
920 | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title