Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD4 Rat anti-Mouse, APC-Alexa Fluor® 750, Clone: RM4-5, Invitrogen™ Non-distribution product as customer accommodation.

Rat Monoclonal Antibody

Manufacturer:  Invitrogen MCD0427

Catalog No. 50-113-7466


Add to cart

Description

Description

The CD4 antigen is involved in the recognition of MHC class II molecules and is a co-receptor for HIV. CD4 is primarily expressed in a subset of T-lymphocytes, also referred to as T helper cells, but may also be expressed by other cells in the immune system, such as monocytes, macrophages, and dendritic cells. At the tissue level, CD4 expression may be detected in thymus, lymph nodes, tonsils, and spleen, and also in specific regions of the brain, gut, and other non-lymphoid tissues. CD4 functions to initiate or augment the early phase of T-cell activation through its association with the T-cell receptor complex and protein tyrosine kinase, Lck. It may also function as an important mediator of direct neuronal damage in infectious and immune-mediated diseases of the central nervous system. Multiple alternatively spliced transcripts have been identified in this gene [RefSeq, July 2017].
Specifications

Specifications

CD4
Monoclonal
0.1 mg/mL
PBS with 4mg/mL BSA, sucrose and 0.1% sodium azide
P06332
CD4 antigen p55, CD4 receptor, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4
Rat
IgG2a
1 mL
4° C
Primary
12504
Flow Cytometry
RM4-5
APC-Alexa Fluor® 750
Cd4
Liquid
Cd4
Mouse CD4
Purified
RUO
Antibody
Monoclonal
Mouse
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit