Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD4 Rat anti-Mouse, PerCP, Clone: GK1.5, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP225191PCP

Catalog No. NP225191PCP


Add to cart

Description

Description

CD4 Monoclonal antibody specifically detects CD4 in Mouse samples. It is validated for Flow Cytometry.
Specifications

Specifications

CD4
GK1.5
Flow Cytometry, Flow (Cell Surface)
CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4
Rat
IgG Kappa
Store at 4°C in the dark.
Monoclonal
Mouse
Flow Cytometry
PerCP
PBS with 0.05% sodium azide
CD4
The mouse cytotoxic T lymphocyte line V4 was used as the immunogen for the CD4 antibody.
0.1 mL
Primary
920
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit