Learn More
CD40 Ligand/TNFSF5 Rabbit anti-Human, Alexa Fluor™ 350, Clone: hu5c8 (Ruplizumab), Novus Biologicals™
Rabbit Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP252678AF350
Catalog No. NP22678A30
Add to cart
Description
Description
CD40 Ligand/TNFSF5 Monoclonal antibody specifically detects CD40 Ligand/TNFSF5 in Human samples. It is validated for Western Blot,Flow Cytometry.
Specifications
Specifications
CD40 Ligand/TNFSF5
hu5c8 (Ruplizumab)
Western Blot, Flow Cytometry
CD154, CD154 antigen, CD40 antigen ligand, CD40 ligand, CD40-L, CD40LIGM, gp39, hCD40L, HIGM1, T-B cell-activating molecule, T-BAM, T-cell antigen Gp39, TNF-related activation protein, TNFSF5IMD3, TRAPtumor necrosis factor (ligand) superfamily, member 5 (hyper-IgM syndrome), tumor necrosis factor (ligand) superfamily member 5, Tumor necrosis factor ligand superfamily member 5
Rabbit
IgG Kappa
Store at 4°C in the dark.
Monoclonal
The antibody binds to CD40L with an IC50 of 0.845 ug/ml.
Flow Cytometry, Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
CD40LG
Human CD40L
0.1 mL
Primary
959
Human