Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD40/TNFRSF5, Human anti-Human,Cynomolgus Monkey, Clone: 5D12, Novus Biologicals™

Human Monoclonal Antibody

$135.50 - $419.00

Specifications

Antigen CD40/TNFRSF5
Clone 5D12
Applications Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Blocking Assay
Host Species Human
Target Species Human, Cynomolgus Monkey
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB016157
View Documents
Novus Biologicals
NBP2811040.025MG
0.025 mg
Each for $135.50
Add to cart
 
NB016158
View Documents
Novus Biologicals
NBP2811040.2MG
0.2 mg
Each for $419.00
Add to cart
 
Description

Description

CD40/TNFRSF5 Monoclonal antibody specifically detects CD40/TNFRSF5 in Human, Cynomolgus Monkey Samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Surface Plasmon Resonance, Blocking Assay.
Specifications

Specifications

CD40/TNFRSF5
Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Blocking Assay
Human, Cynomolgus Monkey
Primary
5D12
Human
Monoclonal
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit