Learn More
CD40/TNFRSF5 Mouse anti-Human, Alexa Fluor™ 350, Clone: G28.5, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP252697AF350
Catalog No. NP22697A30
Add to cart
Description
Description
CD40/TNFRSF5 Monoclonal antibody specifically detects CD40/TNFRSF5 in Human samples. It is validated for Flow Cytometry,Immunohistochemistry.
Specifications
Specifications
CD40/TNFRSF5
G28.5
Flow Cytometry, Immunohistochemistry
B cell surface antigen CD40, B-cell surface antigen CD40, Bp50B cell-associated molecule, CD40 antigen, CD40 molecule, TNF receptor superfamily member 5, CD40 type II isoform, CD40L receptor, CDw40, MGC9013, nerve growth factor receptor-related B-lymphocyte activation molecule, p50, TNFRSF5CD40 antigen (TNF receptor superfamily member 5), tumor necrosis factor receptor superfamily member 5, tumor necrosis factor receptor superfamily, member 5
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
48 kD Type I glycoprotein, a member of the TNFR superfamily primarily expressed on normal or malignant B cells or B cell lines, macrophages, follicular dendritic cells, endothelial cells, fibroblasts, and at low levels on plasma cells.
Flow Cytometry, Immunohistochemistry
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
CD40
Human erythrocyte-rosette-negative tonsillar lymphocytes.
0.1 mL
Primary
958
Human