Learn More
CD40/TNFRSF5 Mouse anti-Human, DyLight 350, Clone: LOB7/6, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP139556UV
Catalog No. NBP139556UV
Add to cart
Description
Description
CD40/TNFRSF5 Monoclonal antibody specifically detects Antigen in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin).
Specifications
Specifications
CD40/TNFRSF5
LOB7/6
Flow Cytometry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Purified
CD40
Recognizes human CD40 cell surface antigen, a 48kD glycoprotein expressed by B lymphocytes and weakly by some monocytes. CD40 is involved in the process of B cell selection in germinal centers and is vital in T cell-B cell interactions.
Protein A purified
RUO
Store at 4C in the dark.
Monoclonal
Recognizes human CD40 cell surface antigen, a 48kD glycoprotein expressed by B lymphocytes and weakly by some monocytes. CD40 is involved in the process of B cell selection in germinal centers and is vital in T cell-B cell interactions.
Flow Cytometry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
B cell surface antigen CD40, B-cell surface antigen CD40, Bp50B cell-associated molecule, CD40 antigen, CD40 molecule, TNF receptor superfamily member 5, CD40 type II isoform, CD40L receptor, CDw40, MGC9013, nerve growth factor receptor-related B-lymphocyte activation molecule, p50, TNFRSF5CD40 antigen (TNF receptor superfamily member 5), tumor necrosis factor receptor superfamily member 5, tumor necrosis factor receptor superfamily, member 5
Mouse
IgG2a
0.1 ml
Adaptive Immunity, Apoptosis, Asthma, B Cell Development and Differentiation Markers, Cancer, Hematopoietic Stem Cell Markers, Immunology, Signal Transduction, Stem Cell Markers
Primary
958
Human