Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD40/TNFRSF5 Rabbit anti-Human, Alexa Fluor™ 594, Clone: G28.5, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP252648AF594
Description
CD40/TNFRSF5 Monoclonal specifically detects CD40/TNFRSF5 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry.Specifications
CD40/TNFRSF5 | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
CD40 | |
Human erythrocyte-rosette-negative tonsillar lymphocytes. | |
Primary | |
48 kD Type I glycoprotein, a member of the TNFR superfamily primarily expressed on normal or malignant B cells or B cell lines, macrophages, follicular dendritic cells, endothelial cells, fibroblasts, and at low levels on plasma cells. | |
Store at 4°C in the dark. |
Flow Cytometry, Immunohistochemistry | |
G28.5 | |
Flow Cytometry, Immunohistochemistry | |
B cell surface antigen CD40, B-cell surface antigen CD40, Bp50B cell-associated molecule, CD40 antigen, CD40 molecule, TNF receptor superfamily member 5, CD40 type II isoform, CD40L receptor, CDw40, MGC9013, nerve growth factor receptor-related B-lymphocyte activation molecule, p50, TNFRSF5CD40 antigen (TNF receptor superfamily member 5), tumor necrosis factor receptor superfamily member 5, tumor necrosis factor receptor superfamily, member 5 | |
Rabbit | |
0.1 mL | |
958 | |
Human | |
IgG κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title