Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD40/TNFRSF5, Rat anti-Mouse, Clone: FGK45, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP280074500UG
Description
CD40/TNFRSF5 Monoclonal specifically detects CD40/TNFRSF5 in Mouse samples. It is validated for Flow Cytometry, Functional.Specifications
CD40/TNFRSF5 | |
Monoclonal | |
1 mg/mL | |
Flow Cytometry, Functional | |
B cell surface antigen CD40, B-cell surface antigen CD40, Bp50B cell-associated molecule, CD40 antigen, CD40 molecule, TNF receptor superfamily member 5, CD40 type II isoform, CD40L receptor, CDw40, MGC9013, nerve growth factor receptor-related B-lymphocyte activation molecule, p50, TNFRSF5CD40 antigen (TNF receptor superfamily member 5), tumor necrosis factor receptor superfamily member 5, tumor necrosis factor receptor superfamily, member 5 | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
Recognizes mouse CD40/TNFRSF5. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG2a |
Flow Cytometry, Functional Assay | |
FGK45 | |
Unconjugated | |
PBS | |
CD40 | |
Recombinant mouse CD40/TNFRSF5 fusion protein. | |
500 μg | |
Adaptive Immunity, Apoptosis, Asthma, B Cell Development and Differentiation Markers, Cancer, Hematopoietic Stem Cell Markers, Immunology, Signal Transduction, Stem Cell Markers | |
958 | |
Mouse | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction