Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD40/TNFRSF5, Rat anti-Mouse, Clone: FGK45, Novus Biologicals™

Rat Monoclonal Antibody

$209.00 - $345.50

Specifications

Antigen CD40/TNFRSF5
Clone FGK45
Applications Flow Cytometry, Functional Assay
Host Species Rat
Target Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB013204
View Documents
Novus Biologicals
NBP280075100UG
100 ug
Each for $209.00
Add to cart
 
NB013205
View Documents
Novus Biologicals
NBP280075500UG
500 ug
Each for $345.50
Add to cart
 
Description

Description

CD40/TNFRSF5 Monoclonal antibody specifically detects CD40/TNFRSF5 in Mouse Samples. It is validated for Flow Cytometry, Functional.
Specifications

Specifications

CD40/TNFRSF5
Flow Cytometry, Functional Assay
Mouse
Primary
FGK45
Rat
Monoclonal
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit