Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD40LG, anti-Human, Clone: hu5c8 (Ruplizumab), Abnova™
Monoclonal Antibody
Supplier: Abnova Corporation RAB00090
Specifications
CD40LG | |
Monoclonal | |
Unconjugated | |
In PBS, pH 7.2 (0.02% Proclin 300). | |
CD154/CD40L/HIGM1/IGM/IMD3/T-BAM/TNFSF5/TRAP/gp39/hCD40L | |
Original antibody is raised against native purified human CD40LG. | |
RUO | |
959 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Blocking Assay, Flow Cytometry | |
hu5c8 (Ruplizumab) | |
Blocking Flow Cytometry (1 ug/mL) The optimal working dilution should be determined by the end user. | |
CD40 ligand | |
CD40LG | |
200 μg | |
Primary | |
Human | |
Antibody | |
IgG κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction