Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin beta 3/CD61 Antibody (VIPL2), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP197563JF549
Description
Integrin beta 3/CD61 Monoclonal specifically detects Integrin beta 3/CD61 in Human, Primate samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
Integrin beta 3/CD61 | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ITGB3 | |
The mouse monoclonal antibody VIPL2 recognizes CD61 | |
0.1 mL | |
Primary | |
The mouse monoclonal antibody VIPL2 recognizes CD61, a 90-110 kDa transmembrane glycoprotein of integrin family, expressed on platelets, megacaryocytes, osteoclasts, endothelial cells and other cell types, including leucocytes and smooth muscle cells. | |
Store at 4°C in the dark. | |
IgG1 κ |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
VIPL2 | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen | |
CD61, CD61 antigen, GP3Aplatelet glycoprotein IIIa, GPIIIaintegrin beta-3, integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61), ITG B3, Platelet membrane glycoprotein IIIa | |
Mouse | |
Protein A purified | |
Angiogenesis, Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Extracellular Matrix, Immunology, Signal Transduction, Stem Cell Markers | |
3690 | |
Human, Primate | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction