Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD42b, PE, clone: MB45, Invitrogen™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MHCD42B04

Catalog No. 50-112-1410


Add to cart

Description

Description

Single-chain membrane glycoprotein that forms a non-covalent complex with GPIb. (MW 23kDa) Reactivity with resting and activated platelets, weakly on monocytes, megakaryocytes and attachment site for the platelet plasma membrane to the submembrane cytoskeleton. GPIb/IX complex, functions as the receptor for ristocetin-induced binding of von Willebrand factor and as the von Willebrand factor-depend adhesion receptor.

R-phycoerythrin (PE) is a stable and highly soluble phycobiliprotein which provides maximal absorbance and fluorescence without susceptibility to internal or external fluorescence quenching, thus providing an exceptional quantum yields and molar extinction coefficients.
Specifications

Specifications

CD42b
Monoclonal
PE
GP1BA
Liquid
GP1BA
Human CD42b
Purified
ASR, ASR/IVD
Antibody
Monoclonal
Human
Flow Cytometry
MB45
PBS with BSA, sucrose and 0.1% sodium azide
P07359
CD42a, GPIX, GP-IX, glycoprotein 9, platelet glycoprotein IX, Platelet GPIX
Mouse
IgG1
0.5mL
Store at 2 - 8 °C.
Primary
2811
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit