Learn More
CD43 Rat anti-Mouse, BB700, Clone: S7, BD Horizon™
Rat Monoclonal Antibody
Manufacturer:  BD Biosciences 566509
Catalog No. BDB566509
Add to cart
Description
Description
CD43 Monoclonal antibody specifically detects CD43 in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD43
S7
BB700
Rat
25 μg
Refrigerate (2 to 8°C)
Monoclonal
Flow Cytometry
0.2 mg/mL
Purified
IgG2a, κ
RUO
Primary
Mouse