Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD43/Sialophorin Mouse anti-Human, PE/Atto594, Clone: MEM-59, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP262227PEATT594

Catalog No. N262227P594


Add to cart

Description

Description

CD43/Sialophorin Monoclonal antibody specifically detects CD43/Sialophorin in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

CD43/Sialophorin
MEM-59
Flow Cytometry
Conjugated, Purified
SPN
Human T lymphocytes.
Protein A purified
B Cell Development and Differentiation Markers, Immunology
Primary
6693.0
Human
Flow Cytometry
PE/ATTO 594
PBS with 0.05% Sodium Azide
CD43 antigen, CD43), Galactoglycoprotein, GALGP, Leukocyte sialoglycoprotein, Sialophorin, sialophorin (gpL115, leukosialin, CD43)
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
The antibody IN-05 reacts with insulin, one of the major regulatory endocrine hormones of intermediate metabolism, normally secreted by the beta cells (a type of islet cells) of the pancreas; it is also present in tumors of B cell origin such as insulinoma.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit