Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ CD43/T-Cell Marker Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible reults in immunohistochemistry and flow cytometry applications with Thermo Scientific™ CD43/T-Cell Marker Ab-1, Mouse Monoclonal Antibody.

Manufacturer:  Thermo Scientific™ MS146R7

 View more versions of this product

Catalog No. MS146R7

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Ensure accurate, reproducible reults in immunohistochemistry and flow cytometry applications with Thermo Scientific™ CD43/T-Cell Marker Ab-1, Mouse Monoclonal Antibody.

CD43 (leukosialin, sialophorin, or leukocyte sialoglycoprotein) is a cell surface glycoprotein which is expressed on all thymocytes and T-cells. CD43 is involved in activation of T cells, B cells, NK cells, and monocytes

Host Species: Mouse

Clone: DF-T1

Isotype: IgG1

Species Reactivity: Human. Does not react with rat. Others not tested

Epitope: Not determined

Immunogen: Myeloblastic KG1 cells

Molecular Weight: 95/115/135 kDa (depending upon extent of glycosylation)

Positive Control: Tonsil or lymph node

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Flow Cytometry
Specifications

Specifications

CD43/T-Cell Marker Ab-1
Monoclonal
Unconjugated
Prediluted
Myeloblastic KG1 cells
7mL
T-Cell
Primary
Human
Flow Cytometry, Immunohistochemistry (Paraffin)
DF-T1
Ready-to-use for IHC (UVLP)
Murine
IgG1
IVD
Primary
Monoclonal
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit