Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ CD43/T-Cell Marker Ab-3, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry and flow cytometry experiments with Thermo Scientific™ CD43/T-Cell Marker Ab-3, Mouse Monoclonal Antibody.

Manufacturer:  Thermo Scientific™ MS679P0

 View more versions of this product

Catalog No. MS679P0

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Obtain accurate, reproducible results in immunohistochemistry and flow cytometry experiments with Thermo Scientific™ CD43/T-Cell Marker Ab-3, Mouse Monoclonal Antibody.

CD43 (leukosialin, sialophorin, or leukocyte sialoglycoprotein) is a cell surface glycoprotein which is expressed on all thymocytes and T-cells. CD43 is involved in activation of T cells, B cells, NK cells, and monocytes

Host Species: Mouse

Clone: 84-3C1

Isotype: IgG1

Species Reactivity: Human. Does not react with rat. Others not tested.

Epitope: Not determined

Immunogen: Stimulated human leukocytes

Molecular Weight: 95/115/135 kDa (depending upon extent of glycosylation)

Positive Control: Tonsil or lymph node

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

CD43/T-Cell Marker Ab-3
Monoclonal
200μg/mL
Purified Ab with BSA and Azide
Murine
IgG1
RUO
Primary
Monoclonal
Flow Cytometry, Immunohistochemistry (Paraffin)
84-3C1
Unconjugated
Concentrated
Stimulated human leukocytes
100μL
T-Cell
Primary
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit