Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD44 s Pan Specific Alexa Fluor 405-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems FAB7045V100UG
This item is not returnable.
View return policy
Description
CD44 Monoclonal specifically detects CD44 in Human samples. It is validated for Flow Cytometry.Specifications
CD44 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P16070 | |
CD44 | |
Mouse myeloma cell line NS0-derived recombinant human CD44s Gln21-Pro220 Accession # P16070 | |
RUO | |
960 | |
Human | |
IgG2a |
Flow Cytometry | |
691534 | |
Alexa Fluor 405 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
CD44, CD44 antigen, CD44 molecule (Indian blood group), CD44R, CDw44, cell surface glycoprotein CD44, chondroitin sulfate proteoglycan 8, CSPG8, ECMR-III, Epican, Extracellular matrix receptor III, GP90 lymphocyte homing/adhesion receptor, HCAM, HCELL, hematopoietic cell E- and L-selectin ligand, Heparan sulfate proteoglycan, Hermes antigen, homing function and Indian blood group system, HUTCH-I, Hyaluronate receptor, IN, LHR, MC56, MDU2, MDU2CD44 antigen (homing function and Indian blood group system), MDU3, MDU3CDW44, MIC4, MIC4MGC10468, MUTCH-I, Pgp1, PGP-1, PGP-I, Phagocytic glycoprotein 1, Phagocytic glycoprotein I | |
Mouse | |
100 μg | |
Primary | |
Detects human CD44 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant CD44 from mouse, rat, or pig is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?
Provide Content Correction