Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD44 Mouse anti-Human, BB515, Clone: G44-26 (C26), BD Horizon™

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 564583

Catalog No. BDB564583


Add to cart

Description

Description

CD44 Monoclonal antibody specifically detects CD44 in Human samples. It is validated for Flow cytometry.
Specifications

Specifications

CD44
G44-26 (also known as C26)
Vol. Per test: 5μL
Purified
Murine
25 Tests
Cell Biology
Primary
Human
Flow Cytometry
BB515
Aqueous buffered solution containing ≤0.09% sodium azide.
Phagocytic glycoprotein 1; Pgp-1; H-CAM; Hermes; ECMR III; HUTCH-1
Mouse IgG2b, κ
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BB515 under optimum conditions and unconjugated antibody was removed.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Reseach Use Only.