Learn More
CD44 Rabbit anti-Human, Unconjugated, Clone: HCAM/2875R, Abnova™
Rabbit Monoclonal Antibody.
Manufacturer:  Abnova Corporation RAB00673
Catalog No. 89-339-688
Add to cart
Description
Description
CD44 Monoclonal antibody specifically detects CD44 in Human samples. It is validated for IHC-P.
Specifications
Specifications
CD44
HCAM/2875R
CD44 molecule (Indian blood group)
Liquid
Rabbit
IgG
Store at -20 to -80°C
Monoclonal
Human
Immunohistochemistry (Paraffin)
Unconjugated
CD44
CDW44, CSPG8, ECMR-III, HCELL, IN, LHR, MC56, MDU2, MDU3, MGC10468, MIC4, MUTCH-I, Pgp1
Stimulated human leukocytes
100 μg
Primary
960