Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD44v6 Mouse anti-Human, Clone: CD44v6/1246, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB15066

Catalog No. 89-035-086


Add to cart

Description

Description

Mouse monoclonal antibody raised against partial recombinant human CD44v6.

  • High specificity
  • High reactivity
Specifications

Specifications

CD44v6
CD44v6/1246
Mouse monoclonal antibody raised against partial recombinant human CD44v6.
In 10 mM PBS (0.05% BSA, 0.05% sodium azide).
Liquid
CD44
Recombinant protein corresponding to human CD44v6.
Protein A/G purification
RUO
Primary
960
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed)
Unconjugated
Flow Cytometry (0.5-1 ug/106 cells) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
P16070
CDW44|CSPG8|ECMR-III|HCELL|IN|LHR|MC56|MDU2|MDU3|MGC10468|MIC4|MUTCH-I|Pgp1
Mouse
IgG2a, kappa
100 ug
Store at 4°C.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)