Learn More
CD45.1 Monoclonal Antibody (A20), NovaFluor™ Blue 660-40S, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Supplier: LIFE TECHNOLOGIES M022T02B07

Description
The A20 monoclonal antibody reacts with the mouse CD45 molecule, the leukocyte common antigen (LCA) in CD45.1-expressing mouse strains. The strains that express CD45.1 include SJL/J, DA, STS/A and RIII. CD45.1 is expressed by all leukocytes in these strains. Applications Reported: The A20 antibody has been reported for use in flow cytometric analysis. Applications Tested: The A20 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. NovaFluor dyes are not compatible with DNA intercalating viability dyes. Do not use viability dyes such as propidium iodide, 7-actinomycin D (7-AAD) and DAPI. Invitrogen LIVE/DEAD Fixable Dead Cell stains are recommended for use with NovaFluor dyes. Each NovaFluor conjugate or kit is shipped with CellBlox Blocking Buffer. Use this buffer whenever staining with NovaFluor conjugates, including single color compensation controls using cells. Use 5 μL of CellBlox Blocking Buffer per stained cell sample containing 10^3 to 10^8 cells. Excitation: 509 nm; Emission: 665 nm; Laser: 488 nm (Blue) Laser
CD45.1 Monoclonal antibody specifically detects CD45.1 in Mouse samples. It is validated for Flow CytometrySpecifications
CD45.1 | |
Monoclonal | |
0.1 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P06800 | |
Mouse | |
25 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a λ |
Flow Cytometry | |
A20 | |
NovaFluor Blue 660-40S | |
Ptprc | |
B220; Cd45; CD45 antigen; CD45R; GP180; Lca; L-CA; Leukocyte common antigen; loc; LY5; Ly-5; Lymphocyte antigen 5; lymphocyte common antigen; Lyt-4; protein tyrosine phosphatase, receptor type C; protein tyrosine phosphatase, receptor type, C; protein tyrosine phosphatase, receptor type, c polypeptide; PTPRC; Receptor-type tyrosine-protein phosphatase C; RT7; T200; T200 glycoprotein; T200 leukocyte common antigen | |
Affinity chromatography | |
RUO | |
19264 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.