Learn More
CD45.2 Mouse anti-Mouse, BB700, Clone: 104, BD Horizon™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 566496
 View more versions of this product
Catalog No. BDB566496
Add to cart
Description
Description
CD45.2 Monoclonal antibody specifically detects CD45.2 in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD45.2
104
BB700
Mouse
0.1 mg
Refrigerate (2 to 8°C)
Monoclonal
Flow Cytometry
0.2 mg/mL
Purified
IgG2a, κ
RUO
Primary
Mouse