Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD45.2 Mouse anti-Mouse, BB700, Clone: 104, BD Horizon™
Mouse Monoclonal Antibody
$120.00 - $352.00
Specifications
Antigen | CD45.2 |
---|---|
Clone | 104 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
CD45.2 Monoclonal antibody specifically detects CD45.2 in Mouse samples. It is validated for Flow Cytometry.Specifications
CD45.2 | |
0.2 mg/mL | |
Monoclonal | |
Purified | |
RUO | |
IgG2a κ | |
Refrigerate (2 to 8°C) |
104 | |
Flow Cytometry | |
Brilliant Blue 700 (BB700) | |
Mouse | |
Mouse | |
Primary |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title