Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD45 Mouse anti-Human, AF 647, Clone: 2D1, Southern Biotech™

Mouse Monoclonal Antibody
Supplier: Southern Biotechnology Associates 962431
Description
SouthernBiotech anti-human primary antibodies are available in a variety of conjugated formats including Alexa Fluor and Pacific Blue.
Specifications
CD45 | |
Monoclonal | |
Lot specific | |
Supplied in PBS/NaN3 | |
Human peripheral blood mononuclear cells | |
RUO | |
Human | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
2D1 | |
Alexa Fluor 647 | |
Mouse | |
100 Tests | |
Primary | |
Store at 2-8°C; Protect from light |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title