Learn More
CD45 Mouse anti-Human, Alexa Fluor 647, Clone: 998209, R&D Systems
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB14301R100UG
Catalog No. FB14301R100
Add to cart
This item is not returnable. View return policy
Description
Description
CD45 Monoclonal antibody specifically detects CD45 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD45
998209
Flow Cytometry 0.25-1 μg/106 cells
P08575
B220, CD45, CD45 antigen, CD45R, EC 3.1.3.48, LCA, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, PTPRC, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide
Mouse myeloma cell line NS0-derived recombinant human CD45, Thr24-Asn575, Accession No. P08575
Protein A or G purified from hybridoma culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Detects human CD45 in direct ELISAs.
Flow Cytometry
Alexa Fluor 647
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Purified, Supernatant, Conjugated
Mouse
IgG1
100 μg
Primary
5788.0
Human