Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD45 Antibody (YKIX716.13) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody has been used in 5 publications
$204.00 - $482.50
Specifications
Antigen | CD45 |
---|---|
Clone | YKIX716.13 |
Concentration | 1.0 mg/mL |
Dilution | Flow Cytometry 1:50-1:100, Immunoprecipitation 1:10-1:500 |
Applications | Immunoprecipitation, Flow Cytometry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017755
|
Novus Biologicals
NB100659060.025MG |
0.025 mg |
Each for $204.00
|
|
|||||
NB100659061
|
Novus Biologicals
NB100659060.1MG |
0.1 mg |
Each for $482.50
|
|
|||||
Description
CD45 Monoclonal specifically detects CD45 in Canine, Equine samples. It is validated for Flow Cytometry, Immunoprecipitation.Specifications
CD45 | |
1.0 mg/mL | |
Immunoprecipitation, Flow Cytometry | |
Unconjugated | |
Rat | |
Adaptive Immunity, Cell Biology, Cellular Markers, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Microglia Markers, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers | |
Phosphate buffered saline with 0.09% Sodium Azide | |
5788 | |
Canine thymocytes | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
147 kDa |
YKIX716.13 | |
Flow Cytometry 1:50-1:100, Immunoprecipitation 1:10-1:500 | |
Monoclonal | |
Purified | |
RUO | |
Canine | |
B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide | |
PTPRC | |
IgG2b | |
Protein G purified | |
Recognizes canine CD45 also known as leukocyte common antigen lustered as Canine CD45 in the First Canine Leukocyte Antigen Workshop. CD45 is expressed on all leukocytes in canine peripheral blood and may be against CD45RB isoform. This antibody reacts with CD45 on all outbred mongrels and beagles tested. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title