Learn More
CD45 Rat anti-Mouse, Alexa Fluor 405, Clone: 5C16, Novus Biologicals™
Rat Monoclonal Antibody
Supplier:  Novus Biologicals NB11093609AF405
Catalog No. NBT93609405
Add to Cart
Description
Description
CD45 Monoclonal specifically detects CD45 in Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
Specifications
CD45
Monoclonal
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide
Rat
Protein A or G purified
Adaptive Immunity, Cell Biology, Cellular Markers, Cellular Signaling, Cytokine Research, Glia Markers, Hematopoietic Stem Cell Markers, Immunology, Immunophenotyping, Innate Immunity, MAP Kinase Signaling, Mast Cell Markers, Mesenchymal Stem Cell Markers, Microglia Markers, Modulation of DNA Pools, Myeloid Cell Markers, Myeloid derived Suppressor Cell, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers
5788
Mouse
Purified
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
5C16
Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
P06800
PTPRC
Cell lysates of mouse leukocytes
0.1 mL
Primary
This was selected for its ability to detect mouse CD45 positive leukocytes in IHC.
Store at 4C in the dark.
IgG2