Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD45 Rat anti-Mouse, Clone: YW62.3, Novus Biologicals™
Rat Monoclonal Antibody
$176.85 - $436.00
Specifications
Antigen | CD45 |
---|---|
Clone | YW62.3 |
Concentration | 1.0 mg/mL |
Dilution | Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
Applications | Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10065366
|
Novus Biologicals
NB10065366 |
0.125 mg |
Each for $436.00
|
|
NB017714
|
Novus Biologicals
NB100653660.025MG |
0.025 mg |
Each for $176.85
|
|
Description
CD45 Monoclonal specifically detects CD45 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen.Specifications
CD45 | |
1.0 mg/mL | |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen) | |
Unconjugated | |
Rat | |
Adaptive Immunity, Cell Biology, Cellular Markers, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Microglia Markers, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers | |
B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide | |
PTPRC | |
IgG2b | |
Protein G purified | |
Recognizes the murine CD45 cell surface antigen, a 180-220kD glycoprotein expressed by all leucocytes. N.B. Some reactivity with human tissue has been observed. |
YW62.3 | |
Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
5788 | |
Mouse spleen cells. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title