Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD45R (B220) Monoclonal Antibody (RA3-6B2), Brilliant Ultra Violet™ 737, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
$222.00 - $435.65
Specifications
Antigen | CD45R (B220) |
---|---|
Clone | RA3-6B2 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-255-0864
![]() |
Invitrogen
367045280 |
25 μg |
Each for $222.00
|
![]() |
|||||
50-255-0863
![]() |
Invitrogen
367045282 |
100 μg |
Each
for $435.65
Save 3% Reg : |
![]() |
|||||
Description
CD45R (PTPRC) is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. CD45 contains an extracellular domain, a single transmembrane segment and two tandem intracytoplasmic catalytic domains. CD45 is specifically expressed in hematopoietic cells and has been shown to be an essential regulator of T- and B-cell antigen receptor signaling. CD45 glycoprotein is crucial in lymphocyte development and antigen signaling, serving as an important regulator of Src-family kinases. CD45 protein exists in multiple isoforms as a result of alternative splicing, and these isoforms differ in their extracellular domains, but share identical transmembrane and cytoplasmic domains. Further, these CD45 isoforms differ in their ability to translocate into the glycosphingolipid-enriched membrane domains and their expression depends on cell type and physiological state of the cell. In addition to its role in immunoreceptor signaling, CD45 is important in promoting cell survival by modulating integrin-mediated signal transduction pathway and is also involved in DNA fragmentation during apoptosis. CD45RA is an isoform of the CD45 complex and has restricted expression between different subtypes of lymphoid cells. Diseases associated with CD45 dysfunction include Severe Combined Immunodeficiency (T-cell negative, B-Cell Negative) and Hepatitis C.Specifications
CD45R (B220) | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with BSA and 0.09% sodium azide, pH 7.2 | |
B220, Cd45, CD45 antigen, CD45R, GP180, Lca, L-CA, leucocyte common antigen, leukocyte common antigen, leukocyte common antigen A, leukocyte common antigen B, loc, LY5, Ly-5, Lymphocyte antigen 5, lymphocyte common antigen, Lyt-4, membrane tyrosine phosphatase, protein tyrosine phosphatase receptor type C, protein tyrosine phosphatase, receptor type C, protein tyrosine phosphatase, receptor type, C, protein tyrosine phosphatase, receptor type, c polypeptide, Protein tyrosine phosphatase, receptor-type, c polypeptide, Ptprc, Receptor-type tyrosine-protein phosphatase C, RT7, T200, T200 glycoprotein, T200 leukocyte common antigen | |
PTPRC | |
Primary | |
4°, C, store in dark, DO NOT FREEZE! |
RA3-6B2 | |
Flow Cytometry | |
Brilliant Ultraviolet 737 | |
Rat | |
Human, Mouse | |
P06800, P08575 | |
19264, 5788 | |
IgG2a κ | |
Affinity chromatography | |
PTPRC |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title