Learn More
CD45R (B220) Rat anti-Human, Mouse, Clone: RA3-6B2, eBioscience™
Rat Monoclonal Antibody
Manufacturer: Invitrogen 14045286

Description
Description: The RA3-6B2 monoclonal antibody reacts with exon A-restricted isoform of mouse CD45, a 220 kDa surface molecule. CD45R/B220 epitope is mainly expressed by the B cell lineage from early Pro-B to mature B cells. However, some activated T cells, lymphokine activated killer cells (LAK), NK cell progenitors in the bone marrow, and T cells of the lpr/lpr mutant mouse also express this antigen. Applications Reported: The RA3-6B2 antibody has been reported for use in flow cytometric analysis, immunoprecipitation, and immunohistochemical staining of frozen or formalin-fixed paraffin embedded tissue sections. RA3-6B2 has also been reported in functional studies of B cells. (Please use Functional Grade purified RA3-6B2, cat. 16-0452, in functional assays.). Applications Tested: The RA3-6B2 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 1 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. The RA3-6B2 antibody has been tested by immunohistochemistry of formalin-fixed paraffin embedded mouse tissue using low and high pH antigen retrieval and can be used at less than or equal to 5 µg/mL.
The RA3-6B2 antibody has been tested by immunohistochemistry of frozen mouse tissue and can be used at less than or equal to 5 µg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered. CD45R is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. CD45R contains an extracellular domain, a single transmembrane segment and two tandem intracytoplasmic catalytic domains. It is specifically expressed in hematopoietic cells and has been shown to be an essential regulator of T- and B-cell antigen receptor signaling.Specifications
CD45R (B220) | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P08575, P06800 | |
Ptprc | |
Affinity chromatography | |
RUO | |
19264, 5788 | |
Antibody | |
IgG2a κ |
Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation | |
RA3-6B2 | |
Unconjugated | |
PTPRC | |
B220, CD45R, Cd45, Ly-5, Lyt-4, T200, Cluster designation 45R | |
Rat | |
1 mg | |
Primary | |
Human, Mouse | |
Liquid |
Safety and Handling
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
For Research Use Only.