Learn More
CD45R Rat anti-Mouse, Alexa Fluor® 647, Clone: RA3-6B2, Invitrogen™ Non-distribution product as customer accommodation.
Rat Monoclonal Antibody
Manufacturer:  Invitrogen RM2621
Catalog No. 50-113-8170
Add to cart
Description
Description
CD45 (LCA, leukocyte common antigen) is a receptor-type protein tyrosine phosphatase ubiquitously expressed in all nucleated hematopoietic cells, comprising approximately 10% of all surface proteins in lymphocytes. CD45 glycoprotein is crucial in lymphocyte development and antigen signaling, serving as an important regulator of Src-family kinases. CD45 protein exists as multiple isoforms as a result of alternative splicing; these isoforms differ in their extracellular domains, whereas they share identical transmembrane and cytoplasmic domains. These isoforms differ in their ability to translocate into the glycosphingolipid-enriched membrane domains and their expression depends on cell type and physiological state of the cell. Besides the role in immunoreceptor signaling, CD45 is important in promoting cell survival by modulating integrin-mediated signal transduction pathway and is also involved in DNA fragmentation during apoptosis. CD45RA is an isoform of the CD45 complex and has restricted expression between different subtypes of lymphoid cells.
Specifications
Specifications
CD45R
Monoclonal
Alexa Fluor® 647
Ptprc
Liquid
Ptprc
exon A-specific epitope on CD45
Purified
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
RA3-6B2
PBS with 0.1% sodium azide
P06800
B220, CD45R, Cd45, Ly-5, Lyt-4, T200, Cluster designation 45R
Rat
IgG2a, kappa
1 mL
4° C
Primary
19264