Learn More
CD45RA Mouse anti-Human , BB515, Clone: HI100, BD Biosciences™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 564552
Catalog No. BDB564552
Add to cart
Description
Description
CD45RA Monoclonal antibody specifically detects CD45RA in Human samples. It is validated for Flow Cytometry
Specifications
Specifications
CD45RA
HI100
Aqueous buffered solution containing ≤0.09% sodium azide.
CD45R; PTPRC; LCA; Leukocyte common antigen
Mouse IgG2b, κ
RUO
Primary
Human
Flow Cytometry
BB515
Conjugated
Mouse
100 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BB515 under optimum conditions and unconjugated antibody was removed.
Monoclonal