Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD45RA Mouse anti-Human, Clone: HI100 (Biotin), Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB13679

Catalog No. 89-033-744


Add to cart

Description

Description

Mouse monoclonal antibody raised against human CD45RA.

  • High specificity
  • High reactivity
Specifications

Specifications

CD45RA
HI100
Mouse monoclonal antibody raised against human CD45RA.
In PBS, pH 7.4 (protein stabilizer, 0.09% sodium azide).
Liquid
PTPRC
Fresh human lymph node lymphocytes depleted of surface immunoglobulin positive cells.
Protein A/G purification
RUO
Primary
5788
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed)
Biotin
Flow Cytometry (1 ug/106 cells) Immunofluorescence Immunohistochemistry (Frozen sections) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) The optimal working dilution should be determined by the end user.
P08575
B220|CD45|CD45R|GP180|LCA|LY5|T200
Mouse
IgG2b, kappa
100 ug
Store in the dark at 4°C.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)