Learn More
CD45RA Mouse anti-Human, Clone: JS-83, Abnova™
Mouse monoclonal antibody raised against human CD45RA.
Manufacturer:  Abnova Corporation MAB12709
Catalog No. 89-933-828
Add to cart
Description
Description
Specifications
Specifications
CD45RA
JS-83
Mouse monoclonal antibody raised against human CD45RA.
In PBS, 150mM NaCl, pH 7.2 (0.09% sodium azide).
Mouse
IgG1, kappa
50 ug
Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Flow Cytometry, Immunohistochemistry (PFA fixed)
Unconjugated
Flow Cytometry Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (20 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
Huamn CD45RA
Affinity purification
RUO
Primary
Human