Learn More
CD45RA Mouse anti-Human, PE-Texas Red®, Clone: MEM-56, Invitrogen™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MHCD45RA17
Catalog No. 50-112-1428
Add to cart
Description
Description
The R-phycoerythrin (PE)-Texas Red™ conjugate permits simultaneous multicolor labeling and detection of multiple targets with excitation by a single excitation source-the 488 nm line of the argon-ion laser.
CD45RA is an isoform of the CD45 complex and has restricted expression between different subtypes of lymphoid cells. CD45 (LCA, leukocyte common antigen) is a receptor-type protein tyrosine phosphatase ubiquitously expressed in all nucleated hematopoietic cells, comprising approximately 10% of all surface proteins in lymphocytes. CD45 glycoprotein is crucial in lymphocyte development and antigen signaling, serving as an important regulator of Src-family kinases. CD45 protein exists as multiple isoforms as a result of alternative splicing and these isoforms differ in their extracellular domains, however, they share identical transmembrane and cytoplasmic domains. CD45 isoforms differ in their ability to translocate into the glycosphingolipid-enriched membrane domains and their expression depends on cell type and physiological state of the cell. Besides the role in immunoreceptor signaling, CD45 is important in promoting cell survival by modulating integrin-mediated signal transduction pathway and is also involved in DNA fragmentation during apoptosis. CD45 is expressed on cells of the human hematopoietic lineage, with the exception of mature red cells and is not detected on differentiated cells of other tissues. It is likely that CD45 plays an important role in signal transduction, inhibition or upregulation of various immunological functions. Diseases associated with CD45 dysfunction include severe combined immunodeficiency and Hepatitis C.
Specifications
Specifications
CD45RA
Monoclonal
PE-Texas Red™
PTPRC
Liquid
PTPRC
Human CD45RA
Purified
ASR
Antibody
Monoclonal
Human
Flow Cytometry
MEM-56
PBS with 10% sucrose, BSA and 0.1% sodium azide
P08575
B220, CD45, CD45R, GP180, LCA, LY5, T200
Mouse
IgG2b
0.5 mL
4° C, store in dark
Primary
5788