Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD45RA Rat anti-Mouse, BB700, Clone: 14.8, BD Optibuild™

Rat Monoclonal Antibody

Manufacturer:  BD Biosciences 742113

Catalog No. BDB742113


Add to cart

Description

Description

The 14.8 monoclonal antibody specifically recognizes an exon A-dependent epitope of the CD45 protein, which is found at high density on B cells and at low density on peripheral T cytotoxic/suppressor cells and a very small subset of thymocytes. Nearly all B-lineage cells, including B-cell precursors in fetal liver and adult bone marrow and Ig-secreting cells, but not hematopoietic stem cells or myeloid progenitors, have been reported to be detectable by mAb 14.8. CD45 is a member of the Protein Tyrosine Phosphatase (PTP) family: Its intracellular (COOH-terminal) region contains two PTP catalytic domains, and the extracellular region is highly variable due to alternative splicing of exons 4, 5, and 6 (designated A, B, and C, respectively), plus, differing levels of glycosylation. The CD45 isoforms detected in the mouse are cell type-, maturation-, and activation state-specific. The CD45 isoforms play complex roles in T-cell and B-cell antigen receptor signal transduction. mAb 14.8 has been reported to enhance the proliferative effect of PHA on purified spleen T cells, possibly by replacing a signal normally delivered by accessory cells, to enhance isotype switching during in vitro B-cell responses, and to inhibit antigen-induced p21 [ras] activation.

The antibody was conjugated to BD Horizon™ BB700, which is part of the BD Horizon Brilliant™ Blue family of dyes. It is a polymer-based tandem dye developed exclusively by BD Biosciences. With an excitation max of 485 nm and an emission max of 693 nm, BD Horizon BB700 can be excited by the 488 nm laser and detected in a standard PerCP-Cy™5.5 set (eg, 695/40nm filter). This dye provides a much brighter alternative to PerCP-Cy5.5 with less cross laser excitation off the 405 nm and 355 nm lasers.BD Horizon Brilliant™ Blue 700 (BB700) is a dye that was exclusively developed by BD Biosciences as brighter alternative to PerCP-Cy5.5. This dye also has less cross laser excitation off the 405 nm laser, resulting in less spillover into the violet channels compared to PerCP-Cy5.5.  Due to similar excitation and emission properties, BD Horizon BB700 and PerCP-Cy5.5 cannot be used simultaneously.
Specifications

Specifications

CD45RA
0.2mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
Ptprc, CD45R, CD45, LCA, Leukocyte common antigen, Ly-5, Lyt-4
Radiation-induced NZC mouse B lymphoma WEHI-279
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BB700 under optimal conditions that minimize unconjugated dye and antibody.
RUO
Primary
Murine
14.8
BB700
Affinity Purified
Rat
Rat IgG2b, κ
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit