Learn More
CD45RB Mouse anti-Human, APC, Clone: MEM-55, Invitrogen™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen A15702
Catalog No. 50-113-7726
Add to cart
Description
Description
CD45RB is a receptor-type protein tyrosine phosphatase, CD45 glycoprotein. CD45 is crucial in lymphocyte development and antigen signaling, serving as an important regulator of Src-family kinases, promotes cell survival by modulating integrin-mediated signal transduction pathway and is also involved in DNA fragmentation during apoptosis. CD45 isoforms differ in their extracellular domains, but share identical transmembrane and cytoplasmic domains. CD45 isoforms differ in their ability to translocate into the glycosphingolipid-enriched membrane domains and their expression depends on cell type and physiological state of the cell. CD45RB is expressed in microglia and inflammatory cells. In addition to involvement in immunoreceptor signaling, CD45 is important in promoting cell survival by modulating integrin-mediated signal transduction pathway and is involved in DNA fragmentation during apoptosis. CD45 is expressed on cells of the human hematopoietic lineage with the exception of mature red cells.
Specifications
Specifications
CD45RB
Monoclonal
APC
PTPRC
Liquid
PTPRC
Human CD45RB
Purified
RUO
Antibody
Monoclonal
Human
Flow Cytometry
MEM-55
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4
P08575
B220, CD45, CD45R, GP180, LCA, LY5, T200
Mouse
IgG1
100 tests
4° C
Primary
5788