Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD45RB Rat anti-Mouse, Phycoerythrin, Clone: C363.16A, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP127986PE

Catalog No. NBP127986PE


Add to cart

Description

Description

CD45RB Monoclonal antibody specifically detects Antigen in Mouse samples. It is validated for Flow Cytometry, Western Blotting.
Specifications

Specifications

CD45RB
C363.16A
Flow Cytometry
Purified
PTPRC
Cloned mouse Th cell line
Protein A or G purified
RUO
Store at 4C in the dark.
Monoclonal
An exon B-dependent epitope of CD45 glycoprotein
Flow Cytometry
PE
PBS with 0.05% Sodium Azide
B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide
Rat
IgG2a Kappa
0.25 ml
Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Microglia Markers, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers
Primary
5788
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit