Learn More
CD45RO (T-Cell Marker) Ab-1 Mouse Monoclonal Antibody, Epredia™
Obtain accurate, reproducible results in immunohistochemistry and flow cytometry experiments with Epredia™ CD45RO (T-Cell Marker) Ab-1, Mouse Monoclonal Antibody.
Supplier: Epredia MS112P1ABX
Description
CD45RO (an isoform of leukocyte common antigen) is expressed on most thymocytes, monocytes, macrophages and granulocytes. It is not expressed on normal B lymphocytes. Intrathymic T cell maturation progresses from CD45RA-/CD45RO+ to CD45RA+/CD45RO-; while peripheral T cell maturation progresses from CD45RA+/CD45RO- to CD45RA-/CD45RO+.Host Species: Mouse
Clone: UCHL-1
Isotype: IgG2a/κ
Species Reactivity: Human, Rhesus Monkey and Chimpanzee. Does not react with rat. Others not known
Epitope: Not determined
Immunogen: Interleukin-2-dependent human T lymphocytes
Molecular Weight: 180 kDa
Positive Control: Tonsil or lymph node
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Flow Cytometry
Specifications
CD45RO (T-Cell Marker) Ab-1 | |
Monoclonal | |
1mg/mL | |
Mouse | |
100 μL | |
Oncogenesis | |
Human, Non-human Primate | |
IgG2a κ |
Flow Cytometry, Immunohistochemistry (Paraffin) | |
UCHL-1 | |
Unconjugated | |
Interleukin-2-dependent human T lymphocytes | |
IVD | |
Primary | |
Concentrated |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.