Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD46, PerCP-eFluor™ 710, clone: 8E2, eBioscience™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

$118.00 - $300.00

Specifications

Antigen CD46
Clone 8E2
Target Species Human
Conjugate PerCP-eFluor 710
Isotype IgG1, kappa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-113-0753 Non-distribution product as customer accommodation. View Documents Invitrogen
46046941
25 Tests Each for $118.00
Add to cart
 
50-113-0754 Non-distribution product as customer accommodation. View Documents Invitrogen
46046942
100 Tests Each for $300.00
Add to cart
 
Description

Description

CD46 (MCP, membrane cofactor protein) is a multifunctional cell surface transmembrane protein that binds and inactivates C3b and C4b complement fragments, regulates T cell-induced inflammatory responses by either inhibiting (CD46-1 isoform) or increasing (CD46-2 isoform) the contact hypersensitivity reaction. CD46 also serves as a receptor for several human pathogens (both bacteria and viruses), and its ligation alteres T lymphocyte polarization toward antigen-presenting cells or target cells, inhibiting lymphocyte function. CD46 is a protector of placental tissue and is also expressed on the inner acrosomal membrane of spermatozoa.

Description: The 8E2 monoclonal antibody reacts with human CD46 (membrane cofactor protein). CD46 is a transmembrane glycoprotein consisting of four regularly expressed isoforms, giving an electrophoretic profile of two heterogeneous protein species of 51-58kDa (lower band) and 59-68kDa (upper band). CD46 is expressed on the majority of nucleated cells, and functions as a regulator of complement function by binding complement components C3b and C4b, which serves to protect the host from autologous complement attack. CD46 has been called a pathogen magnet as it is known to play a role in the entry of several pathogens including strains of measles virus, human herpes virus type 6, group A streptococci and Neisseria.Cross-linking TCR on naive human peripheral blood CD4+ T cells together with CD46, rather than CD28, induces more robust proliferation than achieved with stimulation by the costimulatory molecule CD28, as well as other qualitative differences. There is evidence to suggest that stimulation of TCR and CD46 activates natural CD4+CD25+ T regs, which are non-proliferating, but produce high amounts of granzyme A, and kill activated CD4+, CD8+, CD14+ monocytes, immature dendritic cells and mature dendritic cells. Applications Reported: This 8E2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 8E2 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 µL (0.25 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. PerCP-eFluor™ 710 emits at 710 nm and is excited with the blue laser (488 nm); it can be used in place of PerCP-Cyanine5.5. We recommend using a 710/50 bandpass filter, however, the 695/40 bandpass filter is an acceptable alternative. Please make sure that your instrument is capable of detecting this fluorochrome.Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 µL cell sample + 100 µL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser.Filtration: 0.2µm post-manufacturing filtered.
Specifications

Specifications

CD46
Human
IgG1, kappa
Monoclonal
Liquid
Antibody
Store at 2-8°C. Do not freeze. Light-sensitive material.
8E2
PerCP-eFluor 710
Flow Cytometry
Primary
Monoclonal
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
RUO

For Research Use Only.

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit