Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ CD47 Ab-2, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunofluorescence, immunoprecipitation and flow cytometry experiments with Thermo Scientific™ CD47 Ab-2, Mouse Monoclonal Antibody.

$597.50 - $763.00

Specifications

Antigen CD47 Ab-2
Clone B6H12.2
Applications Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 5
Catalog Number Mfr. No. Concentration Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Concentration Quantity Price Quantity & Availability  
MS1302P
View Documents
Thermo Scientific™
MS1302P
200μg/mL 1mL Each for $742.50
Add to cart
 
MS1302P1
View Documents
Thermo Scientific™
MS1302P1
200μg/mL 500μL Each for $597.50
Add to cart
 
MS1302P1ABX
View Documents
Thermo Scientific™
MS1302P1ABX
1mg/mL 100μL Each for $638.50
Add to cart
 
MS1302PABX
View Documents
Thermo Scientific™
MS1302PABX
1mg/mL 200μL Each for $763.00
Add to cart
 
MS1302P0
View Documents
Thermo Scientific™
MS1302P0
200μg/mL 100μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Ensure accurate, reproducible results in immunofluorescence, immunoprecipitation and flow cytometry experiments with Thermo Scientific™ CD47 Ab-2, Mouse Monoclonal Antibody.

CD47 antigen, also known as integrin associated-protein (IAP), is expressed on all hematopoietic cells, including leukocytes, platelets and erythrocytes. It is also expressed on epithelial cells, endothelial cells, fibroblasts and many tumor cell lines. CD47 may play a role as a signal transducer in the regulation of cation fluxes across cell membranes and in the chemotactic and adhesive interactions of leukocytes with endothelial cells.

Host Species: Mouse

Clone: B6H12.2

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Ig domain

Immunogen: Intact CD47 purified from placenta

Molecular Weight: 47-52 kDa (reduced) and 45-50.5, 110 kDa (nonreduced)

Positive Control: Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunofluorescence (Methanol fixed only)
  • Immunoprecipitation (Native only)
  • Flow Cytometry
  • Blocks binding of SIRPα
  • Inhibits in assays in which CD47-integrin association is required

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

CD47 Ab-2
Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
Murine
Hematopoietic
Intact CD47 purified from placenta
B6H12.2
Monoclonal
Concentrated
RUO
Primary
IgG2a kappa
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit